Learn More
Electron Microscopy Sciences Anti-Vibration Platform Silent Compressor
SDP
Catalog No. 5034456
Encompass_Preferred
These compact and oil free air compressors are virtually silent (30dB/A) versions and suitable for running several platforms.
Catalog No. 50-344-56 Supplier Electron Microscopy Sciences Supplier No. 6610
These compact and oil free air compressors are virtually silent (30dB/A) versions and suitable for running several platforms.