Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Silent Signals : Tracking Disease Through PCR Wastewater Testing
SDPSCI_ED

Catalog No. S001101 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
25 Students (5 Groups)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S001101 25 Students (5 Groups)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001101 Supplier EDVOTEK™ Supplier No. 958
Add to Cart
Add to Cart

Students will use PCR and electrophoresis to analyze simulated water samples for the presence of bacteria and viruses

  • Students analyze simulated water samples using PCR and electrophoresis to investigate real-world wastewater surveillance.
  • Engaging, hands-on experience with essential biotechnology tools to detect and identify pathogens.
  • Illustrates how wastewater testing provides early detection of outbreaks and supports public health decision-making.
  • This project was made possible by a Science Education Partnership Award (SEPA), Grant Number 2R44GM143977-02, from the National Institute of General Medical Sciences (NIGMS) of the National Institutes of Health (NIH).
  • Group Size: For 25 students working in 5 groups.
  • Kit Includes: Instructions, PCR EdvoBeads™, LyphoPrimer™ Mix, EdvoQuick™ DNA Ladder, DNA Templates, TE Buffer, UltraSpec-Agarose™, Electrophoresis Buffer (50X), SYBR™ Safe Stain, FlashBlue™ DNA Stain, Microcentrifuge Tubes and PCR Tubes.
  • All You Need: DNA Electrophoresis, Thermal Cycler, Micropipettes 5-50 μL, Blue Light Transilluminator, White Light Box (optional-use if staining with FlashBlue™), Microcentrifuge, and Microwave or Hot Plate.
  • Storage: Some Components Require Freezer Storage Upon Receipt.

Specifications

Grade Level High School, Higher Ed
Grade 9+
Class Size 25 Students (5 Groups)
Product Type PCR Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.