Learn More
Cell Signaling Technology Silent Synapses Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502730919
Encompass
The Silent Synapses Antibody Sampler Kit provides an economical means of detecting the activation of AMPA-type glutamate receptors using phospho-specific and control antibodies. Enables the comparison of AMPA receptor expression with other synaptic components, such as the NMDA-type glutamate receptor subunit GluN1 and the synaptic scaffolding protein PSD95. Contains sufficient antibody for western blot experiments.
  • Includes antibodies against total and phospho-specific AMPA receptor 1 (GluA1) at Ser831 and Ser845
  • Features antibodies against AMPA receptor 2 (GluA2), NMDA receptor 1 (GluN1), and PSD95
  • Includes HRP-linked anti-rabbit secondary antibody
  • Validated for western blotting, immunofluorescence, and immunoprecipitation applications
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol and <0.02% sodium azide; store at -20°C
Catalog No. 50-273-0919 Supplier Cell Signaling Technology Supplier No. 82760T
The Silent Synapses Antibody Sampler Kit provides an economical means of detecting the activation of AMPA-type glutamate receptors using phospho-specific and control antibodies. Enables the comparison of AMPA receptor expression with other synaptic components, such as the NMDA-type glutamate receptor subunit GluN1 and the synaptic scaffolding protein PSD95. Contains sufficient antibody for western blot experiments.
  • Includes antibodies against total and phospho-specific AMPA receptor 1 (GluA1) at Ser831 and Ser845
  • Features antibodies against AMPA receptor 2 (GluA2), NMDA receptor 1 (GluN1), and PSD95
  • Includes HRP-linked anti-rabbit secondary antibody
  • Validated for western blotting, immunofluorescence, and immunoprecipitation applications
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol and <0.02% sodium azide; store at -20°C