Learn More
World Precision Instrument SilFlex tubing
SDP
Catalog No. NC9081983
Encompass
The Silflex tubing is one of the most critical components of the NanoFil system. This 35cm long, flexible tubing has a very precise outer diameter for airtight fitting with the syringe. It also has a small inner diameter for minimum dead volume, and is very durable when handled correctly.
Catalog No. NC9081983 Supplier World Precision Instrument Supplier No. SILFLEX2
The Silflex tubing is one of the most critical components of the NanoFil system. This 35cm long, flexible tubing has a very precise outer diameter for airtight fitting with the syringe. It also has a small inner diameter for minimum dead volume, and is very durable when handled correctly.