Learn More
Trajan Scientific SilFlow 3 port Splitter Kit 1/EA
SDP
Catalog No. 5000494534
Encompass_Preferred
SilFlow GC 3 port splitter microchannel device (MCD) 0 25 mm ID only port A 0 25/0 32 mm ID ports B and C 0 25/0 32 mm ID
Catalog No. 50-004-94534 Supplier Trajan Scientific Supplier No. 123725
SilFlow GC 3 port splitter microchannel device (MCD) 0 25 mm ID only port A 0 25/0 32 mm ID ports B and C 0 25/0 32 mm ID