Learn More
Trajan Scientific SilFlow 3 port Splitter Kit 1/EA
SDP
Supplier:  Trajan Scientific 123723
Encompass_Preferred
SilFlow GC 3 port splitter microchannel device (MCD) 1 1 mm OD only port A 1 1 mm OD ports B and C 0 25/0 32 mm ID
Catalog No. 50-004-94308