Learn More
Trajan Scientific SilFlow 4 port Splitter Kit 1/EA
Supplier: Trajan Scientific 123730
SilFlow GC 4 port splitter kit port A 1 1 mm OD ports B C and D 0 25/0 32 mm ID (SilFlow GC 4 port splitter MCD 1 1 mm OD SilFlow ferrules 0 4 mm ID PK10 SilFlow ferrules 0 5 mm ID PK10 SilFlow ferrules 1 1 mm ID PK10 pre-swage tool 0 4 mm pre-swage tool 0 5 mm SilFlow nuts PK5 SilFlow blanking ferrules and pins PK2 SilTite FingerTite tool mounting bracket and screws)