Learn More
Trajan Scientific SilFlow 4 port Splitter Kit 1/EA
SDP
Supplier:  Trajan Scientific 123733
Encompass_Preferred
SilFlow GC 4 port splitter microchannel device only port A 1 1 mm OD ports B C and D 0 25/0 32 mm ID
Catalog No. 50-004-94476