Learn More
Trajan Scientific SilFlow 4 port Splitter Kit 1/EA
SDP
Catalog No. 5000494240
Encompass_Preferred
SilFlow GC 4 port splitter kit port A 0 53 mm ID ports B C and D 0 25/0 32 mm ID (SilFlow GC 4 port splitter MCD 0 53 mm ID SilFlow ferrules 0 4 mm ID PK10 SilFlow ferrules 0 5 mm ID PK10 SilFlow ferrules 0 7 mm ID PK10 pre-swage tool 0 4 mm pre-swage tool 0 5 mm pre-swage tool 0 7 mm SilFlow nuts PK5 SilFlow blanking ferrules and pins PK2 SilTite FingerTite tool mounting bracket and screws)
Catalog No. 50-004-94240 Supplier Trajan Scientific Supplier No. 123731
SilFlow GC 4 port splitter kit port A 0 53 mm ID ports B C and D 0 25/0 32 mm ID (SilFlow GC 4 port splitter MCD 0 53 mm ID SilFlow ferrules 0 4 mm ID PK10 SilFlow ferrules 0 5 mm ID PK10 SilFlow ferrules 0 7 mm ID PK10 pre-swage tool 0 4 mm pre-swage tool 0 5 mm pre-swage tool 0 7 mm SilFlow nuts PK5 SilFlow blanking ferrules and pins PK2 SilTite FingerTite tool mounting bracket and screws)