Learn More
Trajan Scientific SilFlow FingerTite Ferrule Medical 10/PK
SDP
Catalog No. 5000494350
Encompass_Preferred
SilFlow ferrules 0 4 mm ID for 0 36 mm OD tubing PK10
Catalog No. 50-004-94350 Supplier Trajan Scientific Supplier No. 123706
SilFlow ferrules 0 4 mm ID for 0 36 mm OD tubing PK10