Learn More
Trajan Scientific SilFlow FingerTite Ferrule Medical 5/PK
SDP
Catalog No. 5000494436
Encompass_Preferred
SilFlow ferrules 1 1 mm ID for 1 07 mm OD tubing PK5
Catalog No. 50-004-94436 Supplier Trajan Scientific Supplier No. 123705
SilFlow ferrules 1 1 mm ID for 1 07 mm OD tubing PK5