Learn More
Miles Scientific SILICA GEL GF 250UM 20X20CM SCORED & CHANNELED (25 PLATES/BOX)
SDP
Catalog No. NC9543515
Encompass_Preferred
Silica Gel G & GF UNIPLATES are the most widely used silica gel TLC plates. No organic substances are added to these products, thus making them compatible with sulfuric acid charring visualization. These plates are commonly scraped from the glass backing to isolate separated components for other analytical procedures. They are compatible with all organic solvents, however, they are not recommended for use in systems containing greater than 20% water because of the solubility of the gypsum binder in aqueous solutions. (For water tolerance see the GHL & GHLF series of plates.)
Catalog No. NC9543515 Supplier Miles Scientific Supplier No. P02711
Silica Gel G & GF UNIPLATES are the most widely used silica gel TLC plates. No organic substances are added to these products, thus making them compatible with sulfuric acid charring visualization. These plates are commonly scraped from the glass backing to isolate separated components for other analytical procedures. They are compatible with all organic solvents, however, they are not recommended for use in systems containing greater than 20% water because of the solubility of the gypsum binder in aqueous solutions. (For water tolerance see the GHL & GHLF series of plates.)