Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuflow Systems Saint-Gobain Silicone Stopper, 0.315 x 0.473 x 0.788 in, 10/Pack

Catalog No. NC2327724
Encompass_Preferred

Saint-Gobain Sani-Tech silicone stoppers are ultra-pure, platinum-cured silicone closures engineered for laboratory and bioprocess use. They provide reliable sealing, high chemical resistance, and compatibility with common sterilization methods to minimize particulate generation and contamination risk.

  • Material: Ultra-pure platinum-cured silicone.
  • Dimensions: 0.315 x 0.473 x 0.788 in (8.0 x 12.0 x 20.0 mm).
  • Pack quantity: 10 pieces per pack.
  • Temperature range: -94° to 482°F (-70° to 250°C).
  • Sterilization: Autoclavable, radiation up to 2.5 Mrad (25 kGy), ethylene oxide compatible.
  • Applications: Biology, hematology, microbiology, biopharmaceutical assays and processes.
  • Compliance: Meets FDA/BGA standards for laboratory closure components.

Catalog No. NC2327724 Supplier Accuflow Systems Supplier No. D1069807
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Saint-Gobain Sani-Tech silicone stoppers are ultra-pure, platinum-cured silicone closures engineered for laboratory and bioprocess use. They provide reliable sealing, high chemical resistance, and compatibility with common sterilization methods to minimize particulate generation and contamination risk.

  • Material: Ultra-pure platinum-cured silicone.
  • Dimensions: 0.315 x 0.473 x 0.788 in (8.0 x 12.0 x 20.0 mm).
  • Pack quantity: 10 pieces per pack.
  • Temperature range: -94° to 482°F (-70° to 250°C).
  • Sterilization: Autoclavable, radiation up to 2.5 Mrad (25 kGy), ethylene oxide compatible.
  • Applications: Biology, hematology, microbiology, biopharmaceutical assays and processes.
  • Compliance: Meets FDA/BGA standards for laboratory closure components.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.