Learn More
Trajan Scientific SILIA DEACT VIT 2VSD-150 ID
SDP
Catalog No. 502530225
Encompass_Preferred
Trajan Deactivated (Methyl) Fused Silica Tubing, Diameter Inner: 0.005 in., 0.15 mm, Diameter Outer: 0.008 in., 0.22 mm, Overall Length: 6.56 ft., 2 m, Capillary Tubing, For Use With: Ideal for Biotechnology Applications, and Organic and Aqueous Solvents
Catalog No. 50-253-0225 Supplier Trajan Scientific Supplier No. 0624460
Trajan Deactivated (Methyl) Fused Silica Tubing, Diameter Inner: 0.005 in., 0.15 mm, Diameter Outer: 0.008 in., 0.22 mm, Overall Length: 6.56 ft., 2 m, Capillary Tubing, For Use With: Ideal for Biotechnology Applications, and Organic and Aqueous Solvents