Learn More
Trajan Scientific SILIA DEACT VIT 5VSD-170 ID
SDP
Catalog No. 502530222
Encompass_Preferred
Trajan Deactivated (Methyl) Fused Silica Tubing, Diameter Inner: 0.006 in., 0.17 mm, Diameter Outer: 0.01 in., 0.3 mm, Overall Length: 16.4 ft., 5 m, Capillary Tubing, For Use With: Ideal for Biotechnology Applications, and Organic and Aqueous Solvents
Catalog No. 50-253-0222 Supplier Trajan Scientific Supplier No. 062449
Trajan Deactivated (Methyl) Fused Silica Tubing, Diameter Inner: 0.006 in., 0.17 mm, Diameter Outer: 0.01 in., 0.3 mm, Overall Length: 16.4 ft., 5 m, Capillary Tubing, For Use With: Ideal for Biotechnology Applications, and Organic and Aqueous Solvents