Learn More
Helix Medical Inc SILIC TUBING 0.030"X0.065" 50'
Catalog No. NC0581035
Encompass_Preferred
HelixMark® biomedical/pharmaceutical grade platinum-cured silicone tubing — manufactured with Dow Corning® SILASTIC® Q7-4750 material — provides superior flow properties, is ideal for use in transfer operations as well as other applications requiring nonleachable material, and is able to operate at elevated temperatures while resisting sticking and encrustations. Tubing has standard dimensions and tolerance.
Catalog No. NC0581035 Supplier Helix Medical Inc Supplier No. 6001104
HelixMark® biomedical/pharmaceutical grade platinum-cured silicone tubing — manufactured with Dow Corning® SILASTIC® Q7-4750 material — provides superior flow properties, is ideal for use in transfer operations as well as other applications requiring nonleachable material, and is able to operate at elevated temperatures while resisting sticking and encrustations. Tubing has standard dimensions and tolerance.