Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica 100-200μm, 60Å, EcoChrom™, MP Biomedicals™

Catalog No. ICN02760 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
25 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN02760 500 g
ICN02761 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN02760 Supplier MP Biomedicals, Inc Supplier No. 0202760
Add to Cart
Add to Cart

For Chromatography; Chemical Synthesis

TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 g
Content And Storage Store at Room Temperature.
Type Silica
Physical Form Powder
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
Density 2g/cm3 at 20°C
CAS 7631-86-9
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.