Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc SILICA 10UM 1200A 150X4.6MM
SDP

Catalog No. 50874678
Encompass_Preferred

Silica 10Um; 1200A 150 X 4.6mm

Catalog No. 50-874-678 Supplier YMC America, Inc Supplier No. SL12S111546WT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Silica 10Um; 1200A 150 X 4.6mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.