Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc SILICA 10UM 1200A 500X30MM
SDP

Supplier:  YMC America, Inc SL12S115030WT

Encompass

Silica 10Um; 1200A 500 X 30mm

Catalog No. 50-874-686


May include imposed supplier surcharges.
Only null left
Add to Cart
Request a Quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.