Learn More
Specifications
Specifications
| Quantity | 25 kg |
| Content And Storage | Store at Room Temperature. |
| Product Type | Silica |
| Physical Form | Free-flowing Powder |
| MDL Number | MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
| Density | 2g/cm3 at 20°C |
| CAS | 7631-86-9 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.