Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc SILICA 25UM 1200A 500X50MM
SDP

Catalog No. 50874698
Encompass

Silica 25Um; 1200A 500 X 50mm

Catalog No. 50-874-698 Supplier YMC America, Inc Supplier No. SL12S215051DR
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Silica 25Um; 1200A 500 X 50mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.