Learn More
Sigma Aldrich Silica
Catalog No. 9007735G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
5 g
Catalog No. Quantity
90-077-35G 5 g
1 options
Catalog No. 90-077-35G Supplier Sigma Aldrich Supplier No. 9007735G
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Unique physical properties such as high surface area and ordered pore structure make mesoporous silica useful for diverse applications extending from CO2 sequestration to catalysis and heavy metal removal from waste streams. Our pelletized mesoporous materials are easy to handle and suitable for aforementioned applications especially in column format which is often required during continuous processing.
Specifications
Quantity 5 g
Synonym Mesoporous slica; Mesostructured silica