Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silica-Alumina Catalyst Support, Grade 135

Catalog No. 3433581KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-335-81KG 1 kg
34-335-825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 34-335-81KG Supplier Sigma Aldrich Supplier No. 3433581KG
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silica-alumina catalyst support, grade 135 can be used as:A catalyst to synthesize 2,6-bis(imino)pyridine derivatives by reacting with substituted difluoroanilines/ortho-halogen-anilines and 2,6-diacetylpyridine.A catalyst support to prepare Ca or Ni-based catalysts for deoxygenation reaction.A catalyst support in hydrocarbon cracking process and transesterification reactions.

Specifications

CAS 1335-30-4
MDL Number MFCD00077645
Quantity 1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.