Learn More
Hach Company Series 5000 Silica Analyzer Reagent Set for Hach Series 5000/5500sc analyzers (MPN 2824400)
Catalog No. NC2335550 Shop All Hach Company Products
Encompass
Hach Series 5000 Silica Analyzer Reagent Set (MPN 2824400) supplies the reagents required for silica (SiO2) analysis using Hach silica analyzers. The kit contains three pre-measured reagent bottles formulated for the molybdate-based silica method, enabling routine analysis and stable instrument operation.
  • Includes Molybdate 3 reagent, Citric acid/surfactant reagent, and Amino Acid F reagent.
  • Each reagent supplied in 2.9 L bottles for extended runtime.
  • Formulated for the molybdate-based silica analytical method.
  • Reagents supplied ready to use, reducing preparation time.
  • Supports routine silica (SiO2) measurements and instrument operation.
Catalog No. NC2335550 Supplier Hach Company Supplier No. 2824400
Hach Series 5000 Silica Analyzer Reagent Set (MPN 2824400) supplies the reagents required for silica (SiO2) analysis using Hach silica analyzers. The kit contains three pre-measured reagent bottles formulated for the molybdate-based silica method, enabling routine analysis and stable instrument operation.
  • Includes Molybdate 3 reagent, Citric acid/surfactant reagent, and Amino Acid F reagent.
  • Each reagent supplied in 2.9 L bottles for extended runtime.
  • Formulated for the molybdate-based silica analytical method.
  • Reagents supplied ready to use, reducing preparation time.
  • Supports routine silica (SiO2) measurements and instrument operation.