Learn More
TWD Scientific LLC Silica BioXbeads coated with SuperMagOH-500 500nm Beads 1mL (50mg/mL)
SDP
Catalog No. 5000052643 Shop All TWD Scientific LLC Products
Encompass_Preferred
Silica BioXbeads coated with SuperMagOH-500 500nm Beads 1mL (50mg/mL)
Catalog No. 50-000-52643 Supplier TWD Scientific LLC Supplier No. BX70304H1
Silica BioXbeads coated with SuperMagOH-500 500nm Beads 1mL (50mg/mL)