Learn More
TWD Scientific LLC Silica BioXbeads Trial Kit Each kit contains a 1mL (50mg/mL) trial of each of the following BioXbeads BX70301 BX70302 BX70111 and BX70112
SDP
Catalog No. 5000052591 Shop All TWD Scientific LLC Products
Encompass_Preferred
Silica BioXbeads Trial Kit Each kit contains a 1mL (50mg/mL) trial of each of the following BioXbeads BX70301 BX70302 BX70111 and BX70112
Catalog No. 50-000-52591 Supplier TWD Scientific LLC Supplier No. BX70100TRIAL
Silica BioXbeads Trial Kit Each kit contains a 1mL (50mg/mL) trial of each of the following BioXbeads BX70301 BX70302 BX70111 and BX70112