Learn More
TWD Scientific LLC Silica BioXbeads coated with MagCOOH-NS500 500nm Beads 50mL (10mg/mL)
SDP
Catalog No. 5000052651 Shop All TWD Scientific LLC Products
Encompass_Preferred
Silica BioXbeads coated with MagCOOH-NS500 500nm Beads 50mL (10mg/mL)
Catalog No. 50-000-52651 Supplier TWD Scientific LLC Supplier No. BX7010850
Silica BioXbeads coated with MagCOOH-NS500 500nm Beads 50mL (10mg/mL)