Learn More
Epoch Life Science Inc GenCatch™ Anion Exchange DNA Maxi Columns (50/PK)
SDP
Supplier:  Epoch Life Science Inc 2040050
Encompass_Preferred
Ion exchange columns for transfection grade plasmid DNA purification. Anion-exchange technology. 1000 ug (1 mg) plasmid DNA binding capacity. Gravity flow. Compatible with third party buffer sets.
Catalog No. 50-153-8105