Learn More
LUKNOVA INC Luknova Supersep Flash Column, 12G Standard Silica, 40-63 µm (230-400 Mesh), 60 Å, 500 M2/G, Bed Ø 23 X 120 Mm, Loading 0.1% - 10%, 30 Per Pack, 200 Psi, Disposable
Catalog No. NC1388274
Encompass
Luknova Patented Supersep Flash Column Packed With 12 Gram Of Standard Silica Of Particle Size 40-63 µm (230-400 Msesh), Pore Size 60 Å, Surface Area 500 M2/G, Silica Bed Ø 23 X 120 Mm, Sample Loading 0.1% - 10%. These Columns Are Disposable And Each Unit Comes At 30 Columns Per Pack With Maximum Pressure 200 Psi. They Are Designed To Fit Directly In The Teledyne Isco, Biotage Isolera, Interchim Puriflash, Yamazen, Santai, Agela, And Other Flash Systems. Patented Design And Automated Packing Technology Enable Better Quality Consistency, Seperation And Prifiaction Efficiency And Reproducibility.
Catalog No. NC1388274 Supplier LUKNOVA INC Supplier No. FC003012
Luknova Patented Supersep Flash Column Packed With 12 Gram Of Standard Silica Of Particle Size 40-63 µm (230-400 Msesh), Pore Size 60 Å, Surface Area 500 M2/G, Silica Bed Ø 23 X 120 Mm, Sample Loading 0.1% - 10%. These Columns Are Disposable And Each Unit Comes At 30 Columns Per Pack With Maximum Pressure 200 Psi. They Are Designed To Fit Directly In The Teledyne Isco, Biotage Isolera, Interchim Puriflash, Yamazen, Santai, Agela, And Other Flash Systems. Patented Design And Automated Packing Technology Enable Better Quality Consistency, Seperation And Prifiaction Efficiency And Reproducibility.