Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silica, Fumed

Catalog No. S5505100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S5505100G 500 g
S5505500G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S5505100G Supplier Sigma Aldrich Supplier No. S5505100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Fumed Silica (SiO2) has been used as a precursor to synthesize: Chlorosilanes via silanization, which are used to prepare silica/polydimethylsiloxane membranes to study permeation performance. Silicon carbide (SiC) nanoparticles by sol-gel method. Calcium silicate hydrate (C-S-H) by treating with CaO and water via hydrothermal reaction. Si/PtO2 catalyst for the selective conversion of cinnamaldehyde to cinnamic acid by aerobic oxidation.It can be also used in the preparation of: Silica sulfuric acid catalyst used in the synthesis of 3,4-dihydropyrimidin-2(1H)-ones via one-pot three-component Biginelli reaction. HClO4-SiO2 heterogeneous catalyst used in the synthesis of tetrasubstituted imidazoles.Particle sizes are for primary particles, which form branched, chain-like aggregates a few tenths of a micron long.

Specifications

CAS 112945-52-5
Density 2.3 lb/cu.ft at 25°C (bulk density)(lit.)
Molecular Formula O2Si
Linear Formula SiO2
Refractive Index n20/D 1.46
Quantity 500 g
MDL Number MFCD00011232
Synonym Silica; Silicic anhydride; Silicon dioxide; Silicon dioxide amorphous
Molecular Weight (g/mol) 60.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.