Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica gel, 1-2,5mm, contains small amounts of blue indicating granules, Thermo Scientific™

Catalog No. AC269520010
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC269520010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC269520010 Supplier Thermo Scientific Chemicals Supplier No. 269520010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for column chromatography, Indicator
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silica gel, 1-2,5 mm, contains small amounts of blue indicating granules
pH 3 to 7
Color Blue-White
Physical Form Granular
CAS 7631-86-9, 7646-79-9
Density 0.75 to 0.9g/cc
Material Silica Gel
Quantity 1 kg
Molecular Formula O2Si
Loss on Drying 1% max. (160°C)
MDL Number MFCD00011232
Merck Index 15, 8632
Particle Size Sieve residue at 2.5mm: <10%, passing through 1mm: <15%
Synonym Silicon dioxide
Formula Weight 60.08
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.