Learn More
Strem, An Ascensus Company CAS# 63231-67-4. 2kg. Silica gel, 70-200 mesh (TLC). MFCD00011232
SDP
Catalog No. 5090114114
Encompass
CAS# 63231-67-4. 2kg. Silica gel, 70-200 mesh (TLC). MFCD00011232. Molecular Weight: 78.10. Molecular Formula: H2SiO3. Color/form: white pwdr. Strem# 93-1427. http://www.strem.com/catalog/v/93-1427/
Catalog No. 50-901-14114 Supplier Strem, An Ascensus Company Supplier No. 9314272000G
CAS# 63231-67-4. 2kg. Silica gel, 70-200 mesh (TLC). MFCD00011232. Molecular Weight: 78.10. Molecular Formula: H2SiO3. Color/form: white pwdr. Strem# 93-1427. http://www.strem.com/catalog/v/93-1427/