Learn More
Sigma Aldrich Fine Chemicals Biosciences Silica gel, 112926-00-8, MFCD00011232, 100 g
Catalog No. NC1036592
Encompass
High-purity grade (Davisil Grade 636), pore size 60 Å, 35-60 mesh particle size, powder, 99%. Silica gel is a rigid 3D network of colloidal silica. It is exclusively used as a support material for the active titanium(III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation leading to form uniform polymer particles with narrow particle size distribution and high bulk density.
Catalog No. NC1036592 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 236802100G
High-purity grade (Davisil Grade 636), pore size 60 Å, 35-60 mesh particle size, powder, 99%. Silica gel is a rigid 3D network of colloidal silica. It is exclusively used as a support material for the active titanium(III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation leading to form uniform polymer particles with narrow particle size distribution and high bulk density.