Learn More
Strem, An Ascensus Company CAS# 63231-67-4. 500g. Silica gel, 6-16 mesh (indicating). MFCD00011232
SDP
Catalog No. 5090114109
Encompass
CAS# 63231-67-4. 500g. Silica gel, 6-16 mesh (indicating). MFCD00011232. Molecular Weight: 78.10. Molecular Formula: H2SiO3. Color/form: blue xtl. Strem# 93-1423. http://www.strem.com/catalog/v/93-1423/
Catalog No. 50-901-14109 Supplier Strem, An Ascensus Company Supplier No. 931423500G
CAS# 63231-67-4. 500g. Silica gel, 6-16 mesh (indicating). MFCD00011232. Molecular Weight: 78.10. Molecular Formula: H2SiO3. Color/form: blue xtl. Strem# 93-1423. http://www.strem.com/catalog/v/93-1423/