Learn More
Sigma Aldrich Fine Chemicals Biosciences SILICA GEL 60 (0.040-0.063 MM)
Catalog No. NC1643425
Encompass
Silica gel 60 (0.040-0.063 mm)Silica gel 60 (0.040-0.063 mm) for column chromatography (230-400 mesh ASTM). CAS No. 7631-86-9, EC Number 231-545-4., for column chromatography (230-400 mesh ASTM)
Catalog No. NC1643425 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1093851000
Silica gel 60 (0.040-0.063 mm)Silica gel 60 (0.040-0.063 mm) for column chromatography (230-400 mesh ASTM). CAS No. 7631-86-9, EC Number 231-545-4., for column chromatography (230-400 mesh ASTM)