Learn More
Sigma Aldrich Fine Chemicals Biosciences Silica gel technical grade, pore size 60 Ã…, 230-400 mesh particle size, 40-63 mum particle size | 112926-00-8 | MFCD00011232 | 5KG
Catalog No. NC1804691
Encompass
Silica gel technical grade, pore size 60 Ã…, 230-400 mesh particle size, 40-63 mum particle size | 112926-00-8 | MFCD00011232 | 5KG
Catalog No. NC1804691 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 7171855KG
Silica gel technical grade, pore size 60 Ã…, 230-400 mesh particle size, 40-63 mum particle size | 112926-00-8 | MFCD00011232 | 5KG