Learn More
Silica Gel 60, 35 to 70 μm, for column chromatography, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Particle Size | 35 to 70 μm |
| Quantity | 1 kg |
| Product Type | Silica Gel 60 |
| Material | Silica 60 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.