Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica Gel 60 SilicAR, Macron Fine Chemicals™

Catalog No. MKV15325 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
25 kg
Packaging:
POLYSTORMOR
Poly Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-V153-25 25 kg Poly Drum
MKV150-500 500 g POLYSTORMOR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MK-V153-25 Supplier Avantor Supplier No. V15325

Chemical Identifiers

CAS 63231-67-4
Molecular Formula H2O3Si
Molecular Weight (g/mol) 78.098
InChI Key IJKVHSBPTUYDLN-UHFFFAOYSA-N
PubChem CID 14768
IUPAC Name dihydroxy(oxo)silane
SMILES O[Si](=O)O

Specifications

Material Silica Gel
Quantity 25 kg
Packaging Poly Drum
Chemical Name or Material Silica Gel 60, (63-200 Microns)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.