Learn More
Sorbent Technologies Inc Silica Gel, Premium Rf, 60A, 20-45um, 5kg
SDP
Catalog No. 501138845
Encompass_Preferred
Silica Gel, Premium Rf has a porosity of 60A, particle distribution of 20-45um, and a quantity of 5kg. It is used as the stationary phase for Medium Pressure Liquid Chromatography (MPLC). Improve your resolution with this MPLC silica gel whether in the laboratory or in production. When working with difficult separations that give very low Rf-values on initial screenings by TLC and require larger volumes of flash grade silica, you need the improved resolution power that Sorbtech MPLC grades provide. By using a smaller particle size, a higher number of theoretical plates are achieved using the same volume of silica which in turn increases the resolution.
Catalog No. 50-113-8845 Supplier Sorbent Technologies Inc Supplier No. 523005
Silica Gel, Premium Rf has a porosity of 60A, particle distribution of 20-45um, and a quantity of 5kg. It is used as the stationary phase for Medium Pressure Liquid Chromatography (MPLC). Improve your resolution with this MPLC silica gel whether in the laboratory or in production. When working with difficult separations that give very low Rf-values on initial screenings by TLC and require larger volumes of flash grade silica, you need the improved resolution power that Sorbtech MPLC grades provide. By using a smaller particle size, a higher number of theoretical plates are achieved using the same volume of silica which in turn increases the resolution.