Learn More
Sorbent Technologies Inc Silica Gel, Technical Grade, 60A, 40-63um, 2.5kg
SDP
Catalog No. 501138820
Encompass_Preferred
Silica Gel, Technical Grade is an economical choice for general separations and filtration. The silica gel has a porosity of 60A, particle distribution of 40-63um (230x400mesh), and a quantity of 2.5kg. This silica gel gives good chromatographic performance for general, non-critical separations. It is used for the packing of traditional flash chromatography columns.
Catalog No. 50-113-8820 Supplier Sorbent Technologies Inc Supplier No. 409302.5
Silica Gel, Technical Grade is an economical choice for general separations and filtration. The silica gel has a porosity of 60A, particle distribution of 40-63um (230x400mesh), and a quantity of 2.5kg. This silica gel gives good chromatographic performance for general, non-critical separations. It is used for the packing of traditional flash chromatography columns.