Learn More
Sorbent Technologies Inc Silica Gel, Enhanced Grade, 60A, 40-63um, 500g
SDP
Catalog No. 501138822
Encompass_Preferred
Silica Gel, Enhanced Grade is a high purity silica 60 for use in the laboratory or in production. The "tight" particle distribution provides a high resolution chromatographic separation. The silica gel has a porosity of 60A, particle distribution of 40-63um (230x400mesh), and a quantity of 500g. This silica gel is ideal for packing traditional flash chromatography columns. It is commonly used for the purification of natural products, foods, cosmetics, pharmaceuticals, and nutraceuticals.
Catalog No. 50-113-8822 Supplier Sorbent Technologies Inc Supplier No. 40930M05
Silica Gel, Enhanced Grade is a high purity silica 60 for use in the laboratory or in production. The "tight" particle distribution provides a high resolution chromatographic separation. The silica gel has a porosity of 60A, particle distribution of 40-63um (230x400mesh), and a quantity of 500g. This silica gel is ideal for packing traditional flash chromatography columns. It is commonly used for the purification of natural products, foods, cosmetics, pharmaceuticals, and nutraceuticals.