Learn More
Sorbent Technologies Inc Silica Gel, 60A, 63-200um, 1kg
SDP
Catalog No. NC2963152
Encompass_Preferred
Silica Gel, Standard Grade is an affordable silica 60 for use in the laboratory or in production. The silica gel has a porosity of 60A, particle distribution of 63-200um (65x250mesh), and a quantity of 1kg. This silica gel is ideal for packing classical chromatography columns where gravity flow is utilized.
Catalog No. NC2963152 Supplier Sorbent Technologies Inc Supplier No. 409401
Silica Gel, Standard Grade is an affordable silica 60 for use in the laboratory or in production. The silica gel has a porosity of 60A, particle distribution of 63-200um (65x250mesh), and a quantity of 1kg. This silica gel is ideal for packing classical chromatography columns where gravity flow is utilized.