Learn More
Sorbent Technologies Inc Silica Gel, 60A, 63-200um, 5kg
SDP
Catalog No. 501138834
Encompass_Preferred
Silica Gel, Standard Grade is an affordable silica 60 for use in the laboratory or in production. The silica gel has a porosity of 60A, particle distribution of 63-200um (65x250mesh), and a quantity of 5kg. This silica gel is ideal for packing classical chromatography columns where gravity flow is utilized.
Catalog No. 50-113-8834 Supplier Sorbent Technologies Inc Supplier No. 409405
Silica Gel, Standard Grade is an affordable silica 60 for use in the laboratory or in production. The silica gel has a porosity of 60A, particle distribution of 63-200um (65x250mesh), and a quantity of 5kg. This silica gel is ideal for packing classical chromatography columns where gravity flow is utilized.