Learn More
Electron Microscopy Sciences Silica Gel Canister, 100 per pack
SDP
Catalog No. 50930886
Encompass_Preferred
This small container offers more desiccant in less space. It is ship-proof and possesses greater drying capabilities. Designed for use in bottles and small containers. Perfect for protecting against potency loss, odor formation, color changes, caking, mildew and condensation. Capsule shell is made from polyethylene with very fine mesh top and bottom. The canister contains 1 gram silica gel.
Catalog No. 50-930-886 Supplier Electron Microscopy Sciences Supplier No. 7120501
This small container offers more desiccant in less space. It is ship-proof and possesses greater drying capabilities. Designed for use in bottles and small containers. Perfect for protecting against potency loss, odor formation, color changes, caking, mildew and condensation. Capsule shell is made from polyethylene with very fine mesh top and bottom. The canister contains 1 gram silica gel.