Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica Gel Desiccant, Indicating, 6 to 18 Mesh, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02003901 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-901 2.5 kg Poly Bottle
02-003-900 500 g Wide Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-003-901 Supplier Avantor Supplier No. 340105
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 1343-98-2
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 14942
ChEBI CHEBI:26675
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Material Silica Gel
Quantity 2.5 kg
Mesh Size 6 to 18 Mesh
Packaging Poly Bottle
Specific Gravity 1L = 2.10kg
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Silica Gel Desiccant, Indicating
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.