Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica Gel Desiccant , Macron Fine Chemicals™

Catalog No. MK447112 Shop All Avantor Products
Change view
Click to view available options
Quantity:
12 kg
Packaging:
Lined Fiber Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-4471-12 12 kg Lined Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-4471-12 Supplier Avantor Supplier No. 447120

Chemical Identifiers

CAS 1343-98-2
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 14942
ChEBI CHEBI:26675
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Material Silica Gel
Quantity 12 kg
Packaging Lined Fiber Drum
Chemical Name or Material Silica Gel Desiccant, indicating
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.