Learn More
Miles Scientific SILICA GEL COATED ON AL FOIL SHEETS 200UM 20X20CM (25 SHEETS)
SDP
Catalog No. NC0250912
Encompass_Preferred
Analtech’s flexible backed TLC plates offer the same exacting specifications as with the classic glass backed plates assuring batch-to-batch uniformity. Quality is controlled at each step in the manufacturing process — from incoming raw materials to outgoing packaging. Lighter and safer to handle than glass; also unbreakable- Easy to cut with scissors to customize plate size or isolate components - Available with Polyester or Aluminum backing - Attach to reports for permanent record - Minimum storage space required - Inexpensive shipping due to weight
Catalog No. NC0250912 Supplier Miles Scientific Supplier No. P156017
Analtech’s flexible backed TLC plates offer the same exacting specifications as with the classic glass backed plates assuring batch-to-batch uniformity. Quality is controlled at each step in the manufacturing process — from incoming raw materials to outgoing packaging. Lighter and safer to handle than glass; also unbreakable- Easy to cut with scissors to customize plate size or isolate components - Available with Polyester or Aluminum backing - Attach to reports for permanent record - Minimum storage space required - Inexpensive shipping due to weight