Learn More
Miles Scientific Silica Gel GHLF 250um 10x20cm scored (25 plates/box)
SDP
Catalog No. NC9167175
Encompass_Preferred
Plates are 100% H20 resistant and are compatible with aqueous developing solvents. They are less rugged than the plates with organic binder but still provide improved handling and sample application compared to Silica Gel G UNIPLATES and are tough enough to write on with a soft pencil. Hard Layer UNIPLATES with inorganic binder are recommended for all applications including those where visualization occurs involving strong charring procedures or acid sprays or dips. Layers tolerate 100% water. Contains no organic materials, Suitable for aggressive visualization procedures, Standard scoring for 20x20 cm plates yields four 5x20 cm plates, One box of scored 10x20 cm plates yields two hundred 2.5x10 cm plates.
Catalog No. NC9167175 Supplier Miles Scientific Supplier No. P21521
Plates are 100% H20 resistant and are compatible with aqueous developing solvents. They are less rugged than the plates with organic binder but still provide improved handling and sample application compared to Silica Gel G UNIPLATES and are tough enough to write on with a soft pencil. Hard Layer UNIPLATES with inorganic binder are recommended for all applications including those where visualization occurs involving strong charring procedures or acid sprays or dips. Layers tolerate 100% water. Contains no organic materials, Suitable for aggressive visualization procedures, Standard scoring for 20x20 cm plates yields four 5x20 cm plates, One box of scored 10x20 cm plates yields two hundred 2.5x10 cm plates.