Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Silica Gel, High-Purity Grade, 200-400 mesh

Catalog No. 111012597 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2597 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2597 Supplier MilliporeSigma™ Supelco™ Supplier No. 288594500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For column chromatography

High purity grade silica gel is useful for all types of low pressure, medium pressure, and flash column chromatography. It can be applied to the clean-up and purification of a wide range of synthetic and natural compounds. A spherical particle shape results in more homogenous packing characteristics, providing sharper peak adsorption and elution curve.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 200/400 Mesh
Grade High-purity Grade
Packaging Poly Bottle
Quantity 500 g
Surface Area (Metric) 500 m2/g
Physical Form Powder
MDL Number MFCD00011232
CAS 112926-00-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.