Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Silica Gel, High-Purity Grade (Davisil™ Grade 636), 35-60 mesh

Catalog No. 111012610 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2610 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2610 Supplier MilliporeSigma™ Supelco™ Supplier No. 236802100G
Add to Cart
Add to Cart

For column chromatography

High purity grade silica gel is useful for all types of low pressure, medium pressure, and flash column chromatography. It can be applied to the clean-up and purification of a wide range of synthetic and natural compounds. A spherical particle shape results in more homogenous packing characteristics, providing sharper peak adsorption and elution curve.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 35/60 Mesh
Pore Size 60 Å
Grade High-purity Grade (Davisil Grade 636)
Packaging Fiber Drum
pH 6.5 (5% in slurry)
Quantity 100 g
Surface Area (Metric) 480 m2/g
Product Line Davisil
Physical Form Powder
MDL Number MFCD00011232
CAS 112926-00-8
Percent Purity ≥99%
Show More Show Less

Davisil is a registered trademark of W.R. Grace & Co.-Conn.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.